2-Phenylnicotinic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

Synonyms: 2-Phenylnicotinic acid, 2-Phenylpyridine-3-carboxylic acid

P2452-1G P2452-200MG
TCP2452-1GEA 188.49 USD
TCP2452-1G TCP2452-200MG
2-Phenylnicotinic acid ≥98.0% (by GC, titration analysis)
2-Phenylnicotinic acid
Formula: C₁₂H₉NO₂
MW: 199.21 g/mol
MDL Number: MFCD03069417
CAS Number: 33421-39-5
EINECS: 251-510-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=df4a9bcb684f3b6c1ae1ca372ae51c864603063e&vid=5e286c8ade00e2dafdef10e9b7c9ffe43b5f5884&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralisation titration) min. 98.0 %
Melting point 167.0 to 171.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR