trans-4-(trans-4-Propylcyclohexyl)cyclohexanol ≥98.0% (by GC)

Supplier: TCI America

Synonyms: (trans,trans)-4'-Propyl-[1,1'-bi(cyclohexan)]-4-ol

P2434-1G P2434-5G
TCP2434-1GEA 45.32 USD
TCP2434-1G TCP2434-5G
trans-4-(trans-4-Propylcyclohexyl)cyclohexanol ≥98.0% (by GC)
trans-4-(trans-4-Propylcyclohexyl)cyclohexanol
Formula: C₁₅H₂₈O
MW: 224.39 g/mol
MDL Number: MFCD09752068
CAS Number: 82832-72-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fdd78d497f8c6966975b05ca75f4d02c9f7530fc&vid=2fc4833665b8f1652f2329de35880cdb2308cce6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 122.0 to 126.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR