PD 184,352 ≥98.0% (by HPLC)

Supplier: TCI America

Synonyms: 2-(2-Chloro-4-iodoanilino)-N-(cyclopropylmethoxy)-3,4-difluorobenzamide, 2-((2-Chloro-4-iodophenyl)amino)-N-(cyclopropylmethoxy)-3,4-difluorobenzamide, PD 184352, CI-1040

P2174-100MG P2174-25MG
TCP2174-100MGEA 747.78 USD
TCP2174-100MG TCP2174-25MG
PD 184,352 ≥98.0% (by HPLC)
PD 184,352
Formula: C₁₇H₁₄ClF₂IN₂O₂
MW: 478.7 g/mol
MDL Number: MFCD02683961
CAS Number: 212631-79-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c8ee72f20e844b739d65c9f4ca72ceade30710c8&vid=1b4f47c2b8e71705f808866c55acba51d4745b78&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Elemental analysis (Nitrogen) 5.30 to 6.30 %
Melting point 176.0 to 180.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR