Sodium 5-(Trifluoromethyl)pyridine-2-sulfinate ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America
Warning

Synonyms: 5-(Trifluoromethyl)pyridine-2-sulphinic acid sodium salt

O0546-1G O0546-250MG
TCO0546-1GEA 108.15 USD
TCO0546-1G TCO0546-250MG
Sodium 5-(Trifluoromethyl)pyridine-2-sulfinate ≥98.0% (by HPLC, titration analysis)
Sodium 5-(Trifluoromethyl)pyridine-2-sulfinate
Formula: C₆H₃F₃NNaO₂S
MW: 233.14 g/mol
MDL Number: MFCD31442692
CAS Number: 2098851-48-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=df7e52d0ca2ee0805fa88976392f5ce2c2c21482&vid=7619b5156cb260b90e779ac450af764c12df2cf8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR