Orbifloxacin ≥98.0% (by HPLC)

Supplier: TCI America

Synonyms: 1-Cyclopropyl-5,6,8-trifluoro-1,4-dihydro-7-(cis-3,5-dimethyl-1-piperazinyl)-4-oxoquinoline-3-carboxylic acid, 1-Cyclopropyl-7-(cis-3,5-dimethylpiperazin-1-yl)-5,6,8-trifluoro-4-oxo-1,4-dihydroquinoline-3-carboxylic acid

O0516-100MG O0516-500MG
TCO0516-100MGEA 55.62 USD
TCO0516-100MG TCO0516-500MG
Orbifloxacin ≥98.0% (by HPLC)
Orbifloxacin
Formula: C₁₉H₂₀F₃N₃O₃
MDL Number: MFCD00864858
CAS Number: 113617-63-3
Merck Index: 14,06862

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralisation titration) min. 96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR