Ocaperidone ≥98.0% (by HPLC)

Supplier: TCI America

Synonyms: 3-[2-[4-(6-Fluorobenzo[d]isoxazol-3-yl)piperidin-1-yl]ethyl]-2,9-dimethyl-4H-pyrido[1,2-a]pyrimidin-4-one

O0498-100MG O0498-25MG
TCO0498-100MGEA 474.83 USD
TCO0498-100MG TCO0498-25MG
Ocaperidone ≥98.0% (by HPLC)
Ocaperidone
Formula: C₂₄H₂₅FN₄O₂
MW: 420.49 g/mol
MDL Number: MFCD00866668
CAS Number: 129029-23-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dc2d2b674591722b2c3b368f4ae1b4fe027d2381&vid=260ca539f19664bee2f84641c7d6baf9ef54dbf8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Elemental analysis (Nitrogen) 12.80 to 13.80 %
Melting point 180.0 to 184.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR