Norlidocaine hydrochloride ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
|
Warning
|
Synonyms:
Monoethylglycinexylidide hydrochloride, N-(2,6-Dimethylphenyl)-2-(ethylamino)acetamide hydrochloride, Norlidocaine HCl
N1171-100MG
N1171-25MG
TCN1171-100MGEA
503.67
USD
TCN1171-100MG
TCN1171-25MG
Norlidocaine hydrochloride ≥98.0% (by HPLC, titration analysis)
Norlidocaine hydrochloride
Formula:
C₁₂H₁₈N₂O·HCl |
MDL Number:
MFCD21334173 CAS Number: 7729-94-4 |
|
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...