N-[2-(Cyclohexyloxy)-4-nitrophenyl]methanesulfonamide ≥98.0% (by HPLC)
Supplier: TCI America
Synonyms:
N-(2-(Cyclohexyloxy)-4-nitrophenyl)methanesulphonamide, NS-398, N-[2-(Cyclohexyloxy)-4-nitrophenyl]methanesulphonamide
N1152-10MG
N1152-50MG
TCN1152-10MGEA
116.39
USD
TCN1152-10MG
TCN1152-50MG
N-[2-(Cyclohexyloxy)-4-nitrophenyl]methanesulfonamide ≥98.0% (by HPLC)
N-[2-(Cyclohexyloxy)-4-nitrophenyl]methanesulphonamide
Formula:
C₁₃H₁₈N₂O₅S MW: 314.356 g/mol |
MDL Number:
MFCD00882995 CAS Number: 123653-11-2 |
Specification Test Results
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Melting point | 137.0 to 141.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...