(3-(Naphthalen-1-yl)phenyl)boronic acid (contains varying amounts of Anhydride)

Supplier: TCI America
Warning

Synonyms: 3-(1-Naphthyl)phenylboronic acid, 3-(1-Naphthyl)benzeneboronic acid

N1131-1G N1131-5G
TCN1131-1GEA 167.89 USD
TCN1131-1G TCN1131-5G
(3-(Naphthalen-1-yl)phenyl)boronic acid (contains varying amounts of Anhydride)
(3-(Naphthalen-1-yl)phenyl)boronic acid
Formula: C₁₆H₁₃BO₂
MW: 248.088 g/mol
MDL Number: MFCD11045054
CAS Number: 881913-20-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8d62d95d030c46b1394f3f318251478a347e3829&vid=1716d0919b9459b284c1e6efdace7ba3e19fd63d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Light yellow to Light orange powder to crystal
Purity (Neutralisation titration) 97.0 to 108.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR