Netilmicin sulfate ≥98.0% (by HPLC)

Supplier: TCI America
Danger

Synonyms: (2R,3R,4R,5R)-2-(((1S,2S,3R,4S,6R)-4-Amino-3-(((2S,3R)-3-amino-6-(aminomethyl)-3,4-dihydro-2H-pyran-2-yl)oxy)-6-(ethylamino)-2-hydroxycyclohexyl)oxy)-5-methyl-4-(methylamino)tetrahydro-2H-pyran-3,5-diol sulphate(2:5)

N1108-100MG N1108-25MG
TCN1108-100MGEA 49.44 USD
TCN1108-100MG TCN1108-25MG
Netilmicin sulfate ≥98.0% (by HPLC)
Netilmicin sulfate
Formula: C₂₁H₄₁N₅O₇·⁵/₂H₂SO₄
Storage Temperature: Refrigerator
MDL Number: MFCD01311715
CAS Number: 56391-57-2
EINECS: 260-147-3
Merck Index: 14,06479
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c478665b17fe7551ea4ece7c7a90255cc8a44e20&vid=619dd8e9978258f1d3e9ace0740ced739d691930&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (HPLC) (CAD) min. 98.0 area%
Elemental analysis (Nitrogen) 9.20 to 10.20 % (calcd.on dried substance)
Specific rotation [a]20/D +87.0 ~ +96.0 deg (C=3, water, calcd.on dried substance)
Water max. 15.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR