Methyl 1,4-Dioxaspiro[4.5]decane-8-carboxylate

Supplier: TCI America

M3273-1G M3273-5G
TCM3273-1GEA 70.04 USD
TCM3273-1G TCM3273-5G
Methyl 1,4-Dioxaspiro[4.5]decane-8-carboxylate
Methyl 1,4-Dioxaspiro[4.5]decane-8-carboxylate
Formula: C₁₀H₁₆O₄
Boiling Pt: 140 °C
MDL Number: MFCD13195278
CAS Number: 26845-47-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a7b9f6d34fda556a1b898ab096dcde31ac287a69&vid=1fb8516137a3f690f259a62d5d0dbfb38b31c163&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR