7-Methyl-1-tetralone ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 7-Methyl-1,2,3,4-tetrahydronaphthalen-1-one, 7-Methyl-3,4-dihydronaphthalen-1(2H)-one

M3255-1G M3255-5G
TCM3255-1GEA 101.97 USD
TCM3255-1G TCM3255-5G
7-Methyl-1-tetralone ≥98.0% (by GC)
7-Methyl-1-tetralone
Formula: C₁₁H₁₂O
Boiling Pt: 144 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00087600
CAS Number: 22009-37-6
EINECS: 244-717-9

Order Now

Specification Test Results

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR