2-(4-Methoxyphenyl)ethylamine Hydroiodide ≥98.0% (by HPLC)
Supplier: TCI America
|
Warning
|
Synonyms:
4-Methoxyphenethylammonium iodide, 4-(2-Aminoethyl)anisole Hydroiodide, 4-Methoxyphenethylamine Hydroiodide, 2-(4-Methoxyphenyl)ethylammonium iodide
M3240-1G
M3240-5G
TCM3240-1GEA
36.05
USD
TCM3240-1G
TCM3240-5G
2-(4-Methoxyphenyl)ethylamine Hydroiodide ≥98.0% (by HPLC)
2-(4-Methoxyphenyl)ethylamine Hydroiodide
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...