2-(Methoxycarbonyl)phenylboronic acid pinacol ester ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 2-(2-Methoxycarbonylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzoic acid methyll ester, 2-(Methoxycarbonyl)phenylboronic acid Pinacol ester, Methyl 2-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate, 2-Methoxycarbonylphenylboronic acid pinacol ester

M3233-1G M3233-5G
TCM3233-1GEA 30.9 USD
TCM3233-1G TCM3233-5G
2-(Methoxycarbonyl)phenylboronic acid pinacol ester ≥98.0% (by GC)
2-(Methoxycarbonyl)phenylboronic acid pinacol ester
Formula: C₁₄H₁₉BO₄
MW: 262.1093 g/mol
MDL Number: MFCD05663866
CAS Number: 653589-95-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d81bdb0bb22e84b5295de66dac41d0dc68d2f32c&vid=106d4621256a49f41df272421f6967f5edfdf4b2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 43.0 to 47.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR