Methyl-4-bromo-2-fluorobenzoate ≥98.0% (by GC)

Supplier: TCI America
Warning

M3229-25G M3229-5G
TCM3229-25GEA 215.27 USD
TCM3229-25G TCM3229-5G
Methyl-4-bromo-2-fluorobenzoate ≥98.0% (by GC)
Methyl-4-bromo-2-fluorobenzoate
Formula: C₈H₆BrFO₂
MDL Number: MFCD06203731
CAS Number: 179232-29-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=116476fbccf02ad6b2ec43ca53a3c131f38eca6d&vid=215d5a576f37d33ff42939faef0eb248b99cf5d7&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR