Methyl 4-Formyl-3-nitrobenzoate ≥98.0% (by GC)
Supplier: TCI America
Synonyms:
4-Formyl-3-nitrobenzoic acid methyl ester
M3198-1G
M3198-5G
TCM3198-1GEA
29.87
USD
TCM3198-1G
TCM3198-5G
Methyl 4-Formyl-3-nitrobenzoate ≥98.0% (by GC)
Methyl 4-Formyl-3-nitrobenzoate
Formula:
C₉H₇NO₅ MW: 209.16 g/mol |
MDL Number:
MFCD04039968 CAS Number: 153813-69-5 |
Specification Test Results
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 62.0 to 66.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...