5-(4-Methylpiperazin-1-yl)pyridin-2-amine ≥98.0% (by GC)

Supplier: TCI America
Danger

Synonyms: 1-Methyl-4-(6-aminopyridin-3-yl)piperazine

M3176-1G M3176-5G
TCM3176-1GEA 67.98 USD
TCM3176-1G TCM3176-5G
5-(4-Methylpiperazin-1-yl)pyridin-2-amine ≥98.0% (by GC)
5-(4-Methylpiperazin-1-yl)pyridin-2-amine
Formula: C₁₀H₁₆N₄
MW: 192.2608 g/mol
MDL Number: MFCD10009563
CAS Number: 571189-49-6
UN: 3263
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dee1d707cf69c3683e60fe47ab3117cc88cafaff&vid=43134607056ec93ba2a5798cb0a86089d7beded5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Light yellow to Brown powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 95.0 %
Melting point 146.0 to 150.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR