1-Methoxy-4-(trans-4-propylcyclohexyl)benzene ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 1-Methoxy-4-(trans-4-N-propylcyclohexyl)benzene

M3152-25G M3152-5G
TCM3152-25GEA 223.51 USD
TCM3152-25G TCM3152-5G
1-Methoxy-4-(trans-4-propylcyclohexyl)benzene ≥98.0% (by GC)
1-Methoxy-4-(trans-4-propylcyclohexyl)benzene
Formula: C₁₆H₂₄O
MW: 232.37 g/mol
MDL Number: MFCD06658193
CAS Number: 81936-32-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=24f7e51e0781bd08175106bff0515dfda0cff757&vid=e24943e9cbd0e1235648c7705df307d305dff0dd&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Light yellow powder to lump
Purity (GC) min. 98.0 %
Melting point 31.0 to 35.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR