4-(Morpholinomethyl)phenylboronic acid (contains varying amounts of Anhydride)

Supplier: TCI America
Warning

Synonyms: 4-(Morpholinomethyl)benzeneboronic acid, [4-(Morpholin-4-ylmethyl)phenyl]boronic acid

M3128-1G M3128-5G
TCM3128-1GEA 96.82 USD
TCM3128-1G TCM3128-5G
4-(Morpholinomethyl)phenylboronic acid (contains varying amounts of Anhydride)
4-(Morpholinomethyl)phenylboronic acid
Formula: C₁₁H₁₆BNO₃
Melting Pt: 85 °C
MDL Number: MFCD06801707
CAS Number: 279262-23-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7d23c009b50a2b62c300d607d1bdb8eb2a37c232&vid=8f897d94c868bd0c12344a6027f9a1a31c3b3c45&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Light yellow to Light orange powder to crystal
Purity (Neutralisation titration) min. 96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR