4-Morpholinobenzoic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

M3100-1G M3100-5G
TCM3100-1GEA 28.84 USD
TCM3100-1G TCM3100-5G
4-Morpholinobenzoic acid ≥98.0% (by GC, titration analysis)
4-Morpholinobenzoic acid
Formula: C₁₁H₁₃NO₃
Melting Pt: 277 °C
MDL Number: MFCD00588379
CAS Number: 7470-38-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b0a9eb99e1955b91e8ab8c3642f41421a9fc64ef&vid=1c16320344aadf4304e1a283df484de234284115&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralisation titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR