Methyl-2-amino-3-chlorobenzoate ≥98.0% (by GC)

Supplier: TCI America

M3074-1G M3074-5G
TCM3074-1GEA 17.51 USD
TCM3074-1G TCM3074-5G
Methyl-2-amino-3-chlorobenzoate ≥98.0% (by GC)
Methyl 2-amino-3-chlorobenzoate
Formula: C₈H₈ClNO₂
MDL Number: MFCD06797833
CAS Number: 77820-58-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ebb64cd052c323e400769e21bc153eb6b08c04d1&vid=c24d8667016f2a50f75d15dc0a588540918bb9b0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Light yellow powder to lump
Purity (GC) min. 98.0 %
Melting point 35.0 to 39.0 deg-C
Acid Value max. 2.0


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR