Methyl 5-Chlorothiophene-2-carboxylate ≥98.0% (by GC)

Supplier: TCI America

M2942-1G M2942-5G
TCM2942-1GEA 36.05 USD
TCM2942-1G TCM2942-5G
Methyl 5-Chlorothiophene-2-carboxylate ≥98.0% (by GC)
Methyl 5-Chlorothiophene-2-carboxylate
Formula: C₆H₅ClO₂S
Boiling Pt: 226 °C
Melting Pt: 18 °C
MDL Number: MFCD06203698
CAS Number: 35475-03-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=47ff45351debb19c12f2766430649373cda0f510&vid=513363bb017da14b996dd341debfaa6b63d6f0c0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR