Methyl-2-amino-5-fluorobenzoate ≥98.0% (by GC)

Supplier: TCI America
Warning

M2899-25G M2899-5G
TCM2899-25GEA 639.63 USD
TCM2899-25G TCM2899-5G
Methyl-2-amino-5-fluorobenzoate ≥98.0% (by GC)
Methyl-2-amino-5-fluorobenzoate
Formula: C₈H₈FNO₂
Boiling Pt: 102 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03791308
CAS Number: 319-24-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=652da923429b0db922c9d74a4ab00179e0c3adfb&vid=dac12bcda36d8d7b14028022bffbd6bda79e1927&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Yellow to Green powder to lump
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR