Methyl-6-fluorochromane-2-carboxylate ≥98.0% (by GC)

Supplier: TCI America

Synonyms: Methyl 6-Fluorochroman-2-carboxylate, 6-Fluoro-3,4-dihydro-2H-1-benzopyran-2-carboxylic acid methyll ester, Methyl 6-Fluoro-3,4-dihydro-2H-1-benzopyran-2-carboxylate, 6-Fluorochroman-2-carboxylic acid methyll ester

M2879-1G M2879-5G
TCM2879-1GEA 36.05 USD
TCM2879-1G TCM2879-5G
Methyl-6-fluorochromane-2-carboxylate ≥98.0% (by GC)
Methyl-6-fluorochromane-2-carboxylate
Formula: C₁₁H₁₁FO₃
MW: 210.21 g/mol
MDL Number: MFCD09701969
CAS Number: 874649-82-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=be3e03275590a46ca03fc4485561d9dc1de52433&vid=e71b017a94e03bdcf7fee54838d08d12d24314d1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 46.0 to 50.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR