2-Methoxy-5-(trifluoromethyl)pyridine ≥98.0% (by GC)

Supplier: TCI America
Warning

M2832-25G M2832-5G
TCM2832-25GEA 98.88 USD
TCM2832-25G TCM2832-5G
2-Methoxy-5-(trifluoromethyl)pyridine ≥98.0% (by GC)
2-Methoxy-5-(trifluoromethyl)pyridine
Formula: C₇H₆F₃NO
MDL Number: MFCD00153203
CAS Number: 175277-45-9
UN: 1993
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=10955ab68d8db527c3fcc7b0a8c706883d1e47ba&vid=16bed1074d2abfbf7f4e03177351ad2b4b67c123&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR