2-Hydrazino-4-(trifluoromethyl)pyrimidine ≥98.0% (by GC)

Supplier: TCI America
Danger

Synonyms: 2-Hydrazinyl-4-(trifluoromethyl)pyrimidine, [4-(Trifluoromethyl)pyrimidin-2-yl]hydrazine

H1574-1G H1574-5G
TCH1574-1GEA 53.56 USD
TCH1574-1G TCH1574-5G
2-Hydrazino-4-(trifluoromethyl)pyrimidine ≥98.0% (by GC)
2-Hydrazino-4-(trifluoromethyl)pyrimidine
Formula: C₅H₅F₃N₄
MW: 178.118 g/mol
MDL Number: MFCD00115112
CAS Number: 197305-97-8
UN: 2811
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d264bf75eeaefe81b54deab81a721fb3fce39dd6&vid=64724cc630a90caca8e389ac3e509a49e8d5628e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 95.0 %
Melting point 99.0 to 103.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR