4,7-Dichloro-1,10-phenanthroline ≥98.0% (by GC, titration analysis)

Supplier: TCI America

D5690-1G D5690-200MG
TCD5690-1GEA 213.21 USD
TCD5690-1G TCD5690-200MG
4,7-Dichloro-1,10-phenanthroline ≥98.0% (by GC, titration analysis)
4,7-Dichloro-1,10-phenanthroline
Formula: C₁₂H₆Cl₂N₂
MDL Number: MFCD00233882
CAS Number: 5394-23-0

Order Now

Specification Test Results

Appearance White to Almost white powder to crystaline
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR