N,N-Diethyl-2,2,2-trifluoroacetamide ≥98.0% (by GC)

Supplier: TCI America
Warning

D5063-25G D5063-5G
TCD5063-25GEA 433.63 USD
TCD5063-25G TCD5063-5G
N,N-Diethyl-2,2,2-trifluoroacetamide ≥98.0% (by GC)
N,N-Diethyl-2,2,2-trifluoroacetamide
Formula: C₆H₁₀F₃NO
MW: 169.15 g/mol
Boiling Pt: 160 °C
Flash Pt: 43 °C
MDL Number: MFCD00192034
CAS Number: 360-92-9
UN: 1993
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7165c33a2eae9b11e69b9089521da7b6e031aee2&vid=1eda32b6dccdfea4a81f0421c2e3b03d36c0d604&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR