2-Bromobenzo[b]naphtho[2,3-d]furan ≥98.0% (by GC)

Supplier: TCI America
Warning

B5805-1G B5805-5G
TCB5805-1GEA 95.79 USD
TCB5805-1G TCB5805-5G
2-Bromobenzo[b]naphtho[2,3-d]furan ≥98.0% (by GC)
2-Bromobenzo[b]naphtho[2,3-d]furan
Formula: C₁₆H₉BrO
MDL Number: MFCD30187285
CAS Number: 1627917-16-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4d95231c7bf67174cce4a02be2884f177304807d&vid=16f0b9a6281d21723cfe5500f1fac946c0b0eff2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Gray to Brown powder to crystal
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR