α,3-Dibromo-5-fluorotoluene ≥98.0% (by GC)
Supplier: TCI America
|
Danger
|
Synonyms:
3-Bromo-5-fluorobenzyl bromide, 1-Bromo-3-(bromomethyl)-5-fluorobenzene
B5338-25G
B5338-5G
TCB5338-25GEA
323.42
USD
TCB5338-25G
TCB5338-5G
α,3-Dibromo-5-fluorotoluene ≥98.0% (by GC)
α,3-Dibromo-5-fluorotoluene
Formula:
C₇H₅Br₂F |
MDL Number:
MFCD09258959 CAS Number: 216755-57-6 UN: 3261 ADR: 8,III |
|
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 45.0 to 49.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...