5-Azoniaspiro[4.4]nonane tetrafluoroborate ≥98.0% (by total nitrogen basis)

Supplier: TCI America
Danger

A3103-1G A3103-5G
TCA3103-1GEA 84.46 USD
TCA3103-1G TCA3103-5G
5-Azoniaspiro[4.4]nonane tetrafluoroborate ≥98.0% (by total nitrogen basis)
5-Azoniaspiro[4.4]nonane tetrafluoroborate
Formula: C₈H₁₆BF₄N
CAS Number: 129211-47-8
UN: 1759
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=330f1673f9e5ce26bfa7771fac75219cd25b0a04&vid=c9a6b1e841840bda4e45bc3715accac9bdecd456&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (with Total Nitrogen) min. 98.0 %
Melting point 188.0 to 192.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR