WZ 811 ≥97.0% (by titrimetric analysis)

Supplier: TCI America
Warning

Synonyms: N,N'-[1,4-Phenylenebis(methylene)]bis(pyridin-2-amine), N,N'-(1,4-Phenylenebis(methylene))bis(pyridin-2-amine)

W0017-100MG W0017-25MG
TCW0017-100MGEA 180.25 USD
TCW0017-100MG TCW0017-25MG
WZ 811 ≥97.0% (by titrimetric analysis)
WZ 811
Formula: C₁₈H₁₈N₄
Storage Temperature: Refrigerator
MDL Number: MFCD18086914
CAS Number: 55778-02-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d4208292c0e713c6e725a0422fe6583dfce09377&vid=4c0200cc8bbb3ba29d86d080998909d4e14afcf5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 95.0 area%
Purity (Nonaqueous Titration) min. 97.0 %
Melting point 188.0 to 192.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR