Wogonin ≥98.0% (by HPLC)

Supplier: TCI America

Synonyms: 5,7-Dihydroxy-8-methoxy-2-phenyl-4H-chromen-4-one, 5,7-Dihydroxy-8-methoxyflavone

W0010-250MG W0010-50MG
TCW0010-250MGEA 430.54 USD
TCW0010-250MG TCW0010-50MG
Wogonin ≥98.0% (by HPLC)
Wogonin
Formula: C₁₆H₁₂O₅
MW: 284.3 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00017736
CAS Number: 632-85-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=66c0c704c44dd080c9ed95c7affa50fb000f2639&vid=db75b7f19d295002d0c457f62d1e0e4f123f16b1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Light orange to Yellow to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Melting point 205.0 to 209.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR