Poly(2-methyl-2-oxazoline) Azide Terminated n=∼50, ULTROXA®
Supplier: TCI America
Synonyms:
alpha-Methyl-omega-azido-poly[(acetylimino)(1,2-ethanediyl)]
U0134-100MG
U0134-500MG
TCU0134-100MGEA
124.63
USD
TCU0134-100MG
TCU0134-500MG
Poly(2-methyl-2-oxazoline) Azide Terminated n=∼50, ULTROXA®
Poly(2-methyl-2-oxazoline) Azide Terminated
Storage Temperature:
Refrigerator |
CAS Number:
1262975-46-1 |
Specification Test Results
Appearance | White to Orange to Green powder to lump |
Average molecular weight | 4,080 to 6,240 |
Molecular weight distribution | max. 1.2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...