1,3,6,8-Tetramethyl-9H-carbazole ≥98.0% (by GC, total nitrogen)

Supplier: TCI America
Warning

T3882-1G T3882-200MG
TCT3882-1GEA 673.62 USD
TCT3882-1G TCT3882-200MG
1,3,6,8-Tetramethyl-9H-carbazole ≥98.0% (by GC, total nitrogen)
1,3,6,8-Tetramethyl-9H-carbazole
Formula: C₁₆H₁₇N
MDL Number: MFCD18450169
CAS Number: 6558-85-6

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (with Total Nitrogen) min. 98.0 %
Melting point 142.0 to 146.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR