4,4,5,5-Tetramethyl-2-(4-phenoxyphenyl)-1,3,2-dioxaborolane ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenoxybenzene, Phenoxyphenyl-4-boronic acid Pinacol ester

T3831-1G T3831-5G
TCT3831-1GEA 33.99 USD
TCT3831-1G TCT3831-5G
4,4,5,5-Tetramethyl-2-(4-phenoxyphenyl)-1,3,2-dioxaborolane ≥98.0% (by GC, titration analysis)
4,4,5,5-Tetramethyl-2-(4-phenoxyphenyl)-1,3,2-dioxaborolane
Formula: C₁₈H₂₁BO₃
MDL Number: MFCD06795691
CAS Number: 269410-26-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=93e40767cfe33f28059bb8de495d288a7d169ba5&vid=95f617362734ae0e6335bbed6a92a1bfdc20f4c8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 52.0 to 56.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR