3',4',5'-Trifluoroacetophenone ≥98.0% (by GC)

Supplier: TCI America
Warning

Synonyms: 1-(3,4,5-Trifluorophenyl)ethan-1-one

T3825-25G T3825-5G
TCT3825-25GEA 195.7 USD
TCT3825-25G TCT3825-5G
3',4',5'-Trifluoroacetophenone ≥98.0% (by GC)
3',4',5'-Trifluoroacetophenone
Formula: C₈H₅F₃O
MW: 174.12 g/mol
MDL Number: MFCD00083533
CAS Number: 220141-73-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b69015cfa827812e0f112e11fd6b8abe890cf78a&vid=2efa81470c0a36413d5169cfa680af0a1f262672&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR