2,4,8,10-Tetraoxaspiro[5.5]undecane-3,9-diylbis(2-methylpropane-2,1-diyl) bis[3-[3-(tert-butyl)-4-hydroxy-5-methylphenyl]propanoate] ≥95.0% (by GC)

Supplier: TCI America
Warning

T3804-25G T3804-5G
TCT3804-25GEA 42.23 USD
TCT3804-25G TCT3804-5G
2,4,8,10-Tetraoxaspiro[5.5]undecane-3,9-diylbis(2-methylpropane-2,1-diyl) bis[3-[3-(tert-butyl)-4-hydroxy-5-methylphenyl]propanoate] ≥95.0% (by GC)
2,4,8,10-Tetraoxaspiro[5.5]undecane-3,9-diylbis(2-methylpropane-2,1-diyl) bis[3-[3-(tert-butyl)-4-hydroxy-5-methylphenyl]propanoate]
Formula: C₄₃H₆₄O₁₀
MDL Number: MFCD09838745
CAS Number: 90498-90-1
EINECS: 410-730-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=38c368bf92860bd95e86e9fbf3258b5421a4f923&vid=041835a33b4ba3fc5c4b002ca563a52465b3c638&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 95.0 %
Melting point 124.0 to 128.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR