Tris(4-bromophenyl)amine ≥99.0% (by GC), purified by sublimation

Supplier: TCI America
Warning

T3777-1G T3777-200MG
TCT3777-1GEA 217.33 USD
TCT3777-1G TCT3777-200MG
Tris(4-bromophenyl)amine ≥99.0% (by GC), purified by sublimation
Tris(4-bromophenyl)amine
Formula: C₁₈H₁₂Br₃N
MW: 482.01 g/mol
MDL Number: MFCD00009665
CAS Number: 4316-58-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7c59b271d6e599a418af571c024fe45cc83a762b&vid=dfba871264abbd1e325df5ef31c0cee7ab0a46db&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 99.0 %
Melting point 144.0 to 147.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR