2,3,6-Trichlorobenzoic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

Synonyms: 2,3,6-TBA, 2,3,6-trichlorobenzoic acid

T3758-1G T3758-5G
TCT3758-1GEA 29.87 USD
TCT3758-1G TCT3758-5G
2,3,6-Trichlorobenzoic acid ≥98.0% (by GC, titration analysis)
2,3,6-Trichlorobenzoic acid
Formula: C₇H₃Cl₃O₂
MW: 225.46 g/mol
MDL Number: MFCD00053310
CAS Number: 50-31-7
EINECS: 200-026-4
UN: 3077
ADR: 9,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=41fa4090f8449912d26511b547e7ee53ecfb3b4f&vid=c4b68b1f4cb2b8a219721c01e44a2b6d58ce2c3b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 126.0 to 130.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR