4-(Carboxymethyl)phenylboronic acid pinacol ester ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

Synonyms: 2-[(4-Carboxymethyl)phenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-(4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)acetic acid, 2-[4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]acetic acid, 4-(Carboxymethyl)phenylboronic acid Pinacol ester

T3754-1G T3754-200MG
TCT3754-1GEA 113.3 USD
TCT3754-1G TCT3754-200MG
4-(Carboxymethyl)phenylboronic acid pinacol ester ≥98.0% (by GC, titration analysis)
4-(Carboxymethyl)phenylboronic acid pinacol ester
Formula: C₁₄H₁₉BO₄
MDL Number: MFCD04038755
CAS Number: 797755-07-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8d5f3c9369344e2efa26dc9b83481952e3c7b408&vid=8af47c3b1dc81a0a61fc50330a5a8a434fab15fa&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 164.0 to 168.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR