(E,E,E,E)-1,2,4,5-Tetrakis[4-[bis(4-methoxyphenyl)amino]styryl]benzene ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America

Synonyms: (E,E,E,E)-4,4',4'',4'''-[Benzene-1,2,4,5-tetrayltetrakis(ethene-2,1-diyl)]tetrakis[N,N-bis(4-methoxyphenyl)aniline], TOP-HTM-α2

T3722-1G T3722-25G T3722-5G
TCT3722-1GEA 104.03 USD
TCT3722-1G TCT3722-25G TCT3722-5G
(E,E,E,E)-1,2,4,5-Tetrakis[4-[bis(4-methoxyphenyl)amino]styryl]benzene ≥98.0% (by HPLC, total nitrogen)
(E,E,E,E)-1,2,4,5-Tetrakis[4-[bis(4-methoxyphenyl)amino]styryl]benzene
Formula: C₉₄H₈₂N₄O₈
MW: 1395.71 g/mol
CAS Number: 2411528-61-3

Order Now

Specification Test Results

Appearance White to Yellow to Orange powder to crystaline
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR