1-(Trifluoromethyl)cyclobutanecarboxylic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

T3717-1G T3717-200MG
TCT3717-1GEA 217.33 USD
TCT3717-1G TCT3717-200MG
1-(Trifluoromethyl)cyclobutanecarboxylic acid ≥98.0% (by GC, titration analysis)
1-(Trifluoromethyl)cyclobutanecarboxylic acid
Formula: C₆H₇F₃O₂
MW: 168.12 g/mol
Boiling Pt: 76 °C
Flash Pt: 96 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03425197
CAS Number: 277756-45-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7a1bdce1208efeb0e5e7871b718a0b159d5d791c&vid=0e64282e46637f7f0770f2a81e7d90af1a6969c6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR