4,4',4''-(1,3,5-Triazine-2,4,6-triyl)trianiline ≥98.0% (by HPLC)

Supplier: TCI America
Warning

T3695-1G T3695-5G
TCT3695-1GEA 126.69 USD
TCT3695-1G TCT3695-5G
4,4',4''-(1,3,5-Triazine-2,4,6-triyl)trianiline ≥98.0% (by HPLC)
4,4',4''-(1,3,5-Triazine-2,4,6-triyl)trianiline
Formula: C₂₁H₁₈N₆
Melting Pt: 250 °C
MDL Number: MFCD04116311
CAS Number: 14544-47-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=056c489655338a4f55bc9ca1425f344b11117985&vid=28182eec9e9478ea3ae0af947b842a8d3cd8bb62&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Yellow to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR