3-(Trifluoromethyl)cyclohexylamine cis- and trans-mixture ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Danger

Synonyms: 3-(Trifluoromethyl)cyclohexanamine

T3678-1G T3678-5G
TCT3678-1GEA 117.42 USD
TCT3678-1G TCT3678-5G
3-(Trifluoromethyl)cyclohexylamine cis- and trans-mixture ≥98.0% (by GC, titration analysis)
3-(Trifluoromethyl)cyclohexylamine
Formula: C₇H₁₂F₃N
MW: 167.18 g/mol
Boiling Pt: 70 °C
MDL Number: MFCD08444665
CAS Number: 56287-83-3
UN: 2734
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b3cae3196ff6fe3606bb869106e5199121cddf47&vid=5722fed44b2ad5e79f1216755819d067745a2919&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 % (total of isomer)
Purity (Nonaqueous Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR