N,N,N',N'-Tetraphenyl[1,1':4',1'':4'',1'''-quaterphenyl]-4,4'''-diamine ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America

Synonyms: N4,N4,N4''',N4'''-Tetraphenyl-[1,1':4',1'':4'',1'''-quaterphenyl]-4,4'''-diamine

T3657-1G T3657-200MG
TCT3657-1GEA 286.34 USD
TCT3657-1G TCT3657-200MG
N,N,N',N'-Tetraphenyl[1,1':4',1'':4'',1'''-quaterphenyl]-4,4'''-diamine ≥98.0% (by HPLC, total nitrogen)
N,N,N',N'-Tetraphenyl[1,1':4',1'':4'',1'''-quaterphenyl]-4,4'''-diamine
Formula: C₄₈H₃₆N₂
MW: 640.83 g/mol
MDL Number: MFCD09833572
CAS Number: 145898-89-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=90ecca26f8305786a684c6faa9d3ce8049e69da8&vid=6c8b9e761404e38423689aad3333c4095c14b667&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR