Tetrakis(4-nitrophenyl)methane ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America

T3643-1G T3643-5G
TCT3643-1GEA 219.39 USD
TCT3643-1G TCT3643-5G
Tetrakis(4-nitrophenyl)methane ≥98.0% (by HPLC, total nitrogen)
Tetrakis(4-nitrophenyl)methane
Formula: C₂₅H₁₆N₄O₈
MW: 500.42 g/mol
Melting Pt: 339 °C
MDL Number: MFCD12546931
CAS Number: 60532-62-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=60f6e18264e6f656a37c22c9e1e7e8d56bee63c7&vid=3430452a8c472cdd4d259fbd55c42873bf741c95&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR