Trimethylsilyl methacrylate ≥98.0% (by GC) stabilized

Supplier: TCI America
Warning


Stabilization: Stabilized with N-Nitrosodiisopropanolamine

T3580-25ML T3580-5ML
TCT3580-25MLEA 91.67 USD
TCT3580-25ML TCT3580-5ML
Trimethylsilyl methacrylate ≥98.0% (by GC) stabilized
Trimethylsilyl methacrylate
Formula: C₇H₁₄O₂Si
Boiling Pt: 52 °C
Flash Pt: 32 °C
MDL Number: MFCD00053868
CAS Number: 13688-56-7
UN: 1993
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=53b89f10b6460529232b465b23645cdc4b3438e0&vid=f82e881b9f59a496f17c0e1a333a77e731c08da0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR