4,4,5,5-Tetramethyl-2-(1,4-dioxaspiro[4.5]dec-7-en-8-yl)-1,3,2-dioxaborolane ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 8-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1,4-dioxaspiro[4.5]dec-7-ene, 4,4,5,5-Tetramethyl-2-(1,4-dioxaspiro[4.5]dec-7-en-8-yl)-1,3,2-dioxaborolane, 1,4-Dioxaspiro[4,5]dec-7-en-8-boronic acid Pinacol ester

T3566-1G T3566-250MG
TCT3566-1GEA 132.87 USD
TCT3566-1G TCT3566-250MG
4,4,5,5-Tetramethyl-2-(1,4-dioxaspiro[4.5]dec-7-en-8-yl)-1,3,2-dioxaborolane ≥98.0% (by GC, titration analysis)
4,4,5,5-Tetramethyl-2-(1,4-dioxaspiro[4.5]dec-7-en-8-yl)-1,3,2-dioxaborolane
Formula: C₁₄H₂₃BO₄
MW: 266.1410 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD04115656
CAS Number: 680596-79-6

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 68.0 to 72.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR