1,2,3-Trifluoro-5-[trans-4-[2-(trans-4-propylcyclohexyl)ethyl]cyclohexyl]benzene ≥98.0% (by GC)

Supplier: TCI America

T3466-1G T3466-5G
TCT3466-1GEA 91.67 USD
TCT3466-1G TCT3466-5G
1,2,3-Trifluoro-5-[trans-4-[2-(trans-4-propylcyclohexyl)ethyl]cyclohexyl]benzene ≥98.0% (by GC)
1,2,3-Trifluoro-5-[trans-4-[2-(trans-4-propylcyclohexyl)ethyl]cyclohexyl]benzene
Formula: C₂₃H₃₃F₃
MDL Number: MFCD12828865
CAS Number: 131819-24-4

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR